Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193268.1 | NP_001180197.1 | 18 | 2122 | ||
NM_001193269.1 | NP_001180198.1 | 18 | 2026 | |||
NM_012155.2 | NP_036287.1 | 15 | 1487 | |||
NR_034098.1 | 13 | 1207 | ||||
XM_006723125.2 | XP_006723188.1 | 18 | 2203 | |||
XM_006723126.2 | XP_006723189.1 | 18 | 2011 | |||
XM_011526694.2 | XP_011524996.1 | 17 | 2110 | |||
XM_011526695.2 | XP_011524997.1 | 17 | 2071 | |||
XM_011526698.2 | XP_011525000.1 | 15 | 1529 | |||
XM_011526700.2 | XP_011525002.1 | 15 | 1475 | |||
XM_011526701.2 | XP_011525003.1 | 10 | 1078 | |||
XM_017026550.1 | XP_016882039.1 | 14 | 1371 | |||
GenBank mRNA | AB209773.1 | - | 12 | 1223 | ||
AF103939.1 | - | 15 | 1481 | |||
AK026616.1 | - | 13 | 1185 | |||
AK225043.1 | - | 15 | 1473 | |||
AK294668.1 | - | 18 | 1944 | |||
AK295905.1 | - | 18 | 2026 | |||
AK296258.1 | - | 18 | 1955 | |||
AK302961.1 | - | 19 | 2208 | |||
AL096717.1 | - | 15 | 1458 | |||
BC029304.1 | - | 18 | 2189 |