Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PANX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015368.3 | NP_056183.2 | 2 | 620 | ||
XM_017017464.1 | XP_016872953.1 | 2 | 423 | |||
GenBank mRNA | AF398509.1 | - | 2 | 618 | ||
AK074897.1 | - | 2 | 565 | |||
AK313074.1 | - | 2 | 590 | |||
AY359023.1 | - | 2 | 381 | |||
BC016931.1 | - | 2 | 615 | 4390851 | ||
FM201789.1 | - | 2 | 235 |