Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FJX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014344.3 | NP_055159.2 | 1 | 1464 | ||
GenBank mRNA | AJ245599.1 | - | 1 | 1144 | ||
AK026069.1 | - | 1 | 430 | |||
AK315015.1 | - | 1 | 1233 | |||
BC049171.1 | - | 1 | 1210 | 5482332 | ||
BC071898.1 | - | 1 | 1210 | 5470218 |