Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199885.1 | NP_001186814.1 | 1 | 336 | ||
NM_016400.3 | NP_057484.3 | 1 | 336 | |||
GenBank mRNA | AF049613.1 | - | 1 | 133 | ||
AF161485.1 | - | 1 | 157 | |||
AK022435.1 | - | 1 | 158 | |||
BC019262.1 | - | 1 | 334 | 3504222 | ||
BQ478474.1 | - | 1 | 134 | |||
BQ772100.1 | - | 4 | 313 |
Gene Symbol: SERF2-C15ORF63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037673.1 | 3 | 779 | |||
GenBank mRNA | AF370428.1 | - | 2 | 4272 | ||
AK000438.1 | - | 3 | 401 | |||
AK094716.1 | - | 2 | 2104 | |||
AK095876.1 | - | 4 | 1777 |