Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199885.1 | NP_001186814.1 | 3 | 438 | ||
NM_016400.3 | NP_057484.3 | 4 | 494 | |||
GenBank mRNA | AF049613.1 | - | 4 | 291 | ||
AK022435.1 | - | 2 | 401 | |||
BC019262.1 | - | 4 | 492 | 3504222 | ||
BQ478474.1 | - | 3 | 236 | |||
BQ772100.1 | - | 1 | 155 |
Gene Symbol: SERF2-C15ORF63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037673.1 | 6 | 937 | |||
GenBank mRNA | AF370428.1 | - | 5 | 4430 | ||
AK000438.1 | - | 6 | 559 | |||
AK094716.1 | - | 5 | 2262 | |||
AK095876.1 | - | 7 | 1935 |