Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPF40B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031698.2 | NP_001026868.2 | 3 | 321 | ||
NM_012272.2 | NP_036404.1 | 2 | 611 | |||
XM_006719324.3 | XP_006719387.1 | 3 | 1403 | |||
XM_006719325.3 | XP_006719388.1 | 3 | 1403 | |||
XM_011538135.2 | XP_011536437.1 | 3 | 1403 | |||
XM_011538136.2 | XP_011536438.1 | 3 | 1403 | |||
XM_011538137.2 | XP_011536439.1 | 2 | 1322 | |||
XM_011538138.2 | XP_011536440.1 | 3 | 1403 | |||
XM_011538139.2 | XP_011536441.1 | 3 | 315 | |||
XM_011538140.2 | XP_011536442.1 | 3 | 315 | |||
XM_011538141.1 | XP_011536443.1 | 2 | 141 | |||
XM_011538143.2 | XP_011536445.1 | 2 | 157 | |||
XM_011538144.2 | XP_011536446.1 | 2 | 141 | |||
XM_017019135.1 | XP_016874624.1 | 2 | 78 | |||
XM_017019136.1 | XP_016874625.1 | 2 | 141 | |||
XM_017019137.1 | XP_016874626.1 | 2 | 141 | |||
XM_017019138.1 | XP_016874627.1 | 3 | 1403 | |||
GenBank mRNA | AK123353.1 | - | 2 | 84 | ||
AL834216.1 | - | 2 | 611 | |||
BC050398.2 | - | 3 | 157 | |||
BC067364.1 | - | 2 | 142 | 6644672 |