Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284303.1 | NP_001271232.1 | 1 | 200 | ||
NM_001284304.1 | NP_001271233.1 | 1 | 200 | |||
NM_014346.3 | NP_055161.1 | 1 | 200 | |||
XM_005261496.1 | XP_005261553.1 | 1 | 196 | |||
XM_011530099.1 | XP_011528401.1 | 1 | 196 | |||
XM_011530100.2 | XP_011528402.1 | 1 | 196 | |||
XM_011530101.2 | XP_011528403.1 | 1 | 196 | |||
XM_011530102.2 | XP_011528404.1 | 1 | 196 | |||
XM_017028742.1 | XP_016884231.1 | 1 | 196 | |||
XM_017028743.1 | XP_016884232.1 | 1 | 196 | |||
XM_017028744.1 | XP_016884233.1 | 1 | 130 | |||
XM_017028745.1 | XP_016884234.1 | 1 | 196 | |||
XM_017028746.1 | XP_016884235.1 | 1 | 196 | |||
XM_017028747.1 | XP_016884236.1 | 1 | 196 | |||
GenBank mRNA | AB449908.1 | - | 1 | 30 | ||
AK055591.1 | - | 1 | 188 | |||
AK301445.1 | - | 1 | 178 | |||
AK308039.1 | - | 1 | 127 | |||
AK315779.1 | - | 1 | 162 | |||
BC001292.1 | - | 1 | 133 | 3457393 | ||
BC002743.2 | - | 1 | 136 | 3627634 | ||
BC020976.1 | - | 1 | 135 | 3884347 | ||
BC029897.1 | - | 1 | 165 | 5173045 | ||
CR456412.1 | - | 1 | 51 | |||
DA990679.1 | - | 1 | 196 | |||
HY033225.1 | - | 1 | 201 |