Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSTYK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015375.2 | NP_056190.1 | 9 | 2251 | ||
NM_199462.2 | NP_955749.1 | 9 | 2251 | |||
XM_011509392.2 | XP_011507694.1 | 9 | 2182 | |||
XM_011509393.2 | XP_011507695.1 | 10 | 2145 | |||
XM_011509394.2 | XP_011507696.1 | 8 | 1653 | |||
GenBank mRNA | AB007941.1 | - | 4 | 497 | ||
AK298011.1 | - | 8 | 1807 | |||
AY208850.1 | - | 9 | 2187 | |||
AY429674.1 | - | 9 | 2223 | |||
BC048204.1 | - | 2 | 205 | |||
BC053627.1 | - | 9 | 2214 | 6164875 | ||
BC072406.1 | - | 9 | 2195 | 6138109 | ||
BC117411.1 | - | 9 | 2212 | 40125962 | ||
BC143603.1 | - | 9 | 2211 | 9052116 |