Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139488.1 | NP_001132960.1 | 6 | 1024 | ||
NM_015376.2 | NP_056191.1 | 5 | 582 | |||
NM_170672.2 | NP_733772.1 | 7 | 937 | |||
XM_005264246.3 | XP_005264303.1 | 6 | 1364 | |||
XM_011532746.2 | XP_011531048.1 | 7 | 811 | |||
XM_011532747.2 | XP_011531049.1 | 6 | 826 | |||
XM_011532748.2 | XP_011531050.2 | 6 | 672 | |||
XM_017003759.1 | XP_016859248.1 | 8 | 2287 | |||
XM_017003760.1 | XP_016859249.1 | 7 | 2759 | |||
XM_017003761.1 | XP_016859250.1 | 6 | 689 | |||
XM_017003762.1 | XP_016859251.1 | 7 | 938 | |||
XM_017003763.1 | XP_016859252.1 | 6 | 668 | |||
GenBank mRNA | AB020653.1 | - | 5 | 582 | ||
AK292983.1 | - | 7 | 848 | |||
BC027849.1 | - | 7 | 937 | 5203041 | ||
BX647990.1 | - | 6 | 632 |