Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114090.1 | NP_001107562.1 | 4 | 777 | ||
NM_019850.2 | NP_062824.2 | 6 | 1149 | |||
XM_011510923.2 | XP_011509225.1 | 6 | 1115 | |||
XM_011510924.1 | XP_011509226.1 | 2 | 148 | |||
XM_011510925.1 | XP_011509227.1 | 2 | 152 | |||
GenBank mRNA | AJ238899.1 | - | 3 | 463 | ||
AK293203.1 | - | 2 | 149 | |||
AK297390.1 | - | 4 | 777 | |||
AK298315.1 | - | 6 | 1101 | |||
AK300953.1 | - | 7 | 1053 | |||
BC031573.1 | - | 6 | 1136 | 5186132 | ||
BC073962.1 | - | 4 | 780 |