Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012198.3 | NP_036330.1 | 2 | 275 | ||
XM_005246446.2 | XP_005246503.1 | 2 | 216 | |||
XM_006712398.3 | XP_006712461.1 | 2 | 448 | |||
XM_006712400.3 | XP_006712463.1 | 3 | 526 | |||
XM_006712401.3 | XP_006712464.1 | 2 | 527 | |||
XM_011510926.2 | XP_011509228.1 | 2 | 227 | |||
XM_011510927.2 | XP_011509229.1 | 2 | 222 | |||
XM_017003764.1 | XP_016859253.1 | 3 | 539 | |||
XM_017003765.1 | XP_016859254.1 | 3 | 625 | |||
XM_017003766.1 | XP_016859255.1 | 3 | 422 | |||
XM_017003767.1 | XP_016859256.1 | 2 | 298 | |||
XM_017003768.1 | XP_016859257.1 | 2 | 293 | |||
XM_017003769.1 | XP_016859258.1 | 2 | 448 | |||
GenBank mRNA | AB209768.1 | - | 2 | 164 | ||
AK295413.1 | - | 2 | 208 | |||
AK312349.1 | - | 2 | 224 | |||
BC005214.1 | - | 2 | 198 | 3996889 | ||
M81637.1 | - | 2 | 218 |