Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015380.4 | NP_056195.3 | 11, 12 | 1194 | ||
GenBank mRNA | AF151809.1 | - | 11, 12 | 1106 | ||
AK001087.1 | - | 11, 12 | 1133 | |||
AK097015.1 | - | 4, 5 | 1809 | |||
AK222657.1 | - | 11, 12 | 1068 | |||
AK297770.1 | - | 10, 11 | 877 | |||
AK312773.1 | - | 11, 12 | 1029 | |||
AY189688.1 | - | 11, 12 | 1109 | |||
BC007830.1 | - | 11, 12 | 1101 | 3635175 | ||
BC011681.2 | - | 11, 12 | 1154 | 3010765 | ||
BC015200.1 | - | 11, 12 | 1044 | 4333051 | ||
CR456483.1 | - | 11, 12 | 1006 |