Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005744.3 | NP_005735.2 | 12 | 1733 | ||
XM_011521438.2 | XP_011519740.1 | 11 | 1709 | |||
XM_011521439.2 | XP_011519741.1 | 13 | 1247 | |||
GenBank mRNA | AB014774.1 | - | 3 | 293 | ||
AB209432.1 | - | 13 | 1362 | |||
AF072832.1 | - | 12 | 1419 | |||
AJ009771.1 | - | 5 | 529 | |||
AJ130976.1 | - | 12 | 1137 | |||
AJ243190.1 | - | 12 | 1733 | |||
AK312715.1 | - | 12 | 1732 | |||
BC051877.1 | - | 12 | 1668 | 5742498 |