Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HECTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015382.3 | NP_056197.3 | 32 | 6320 | ||
XM_005267502.2 | XP_005267559.2 | 32 | 6482 | |||
XM_011536619.2 | XP_011534921.1 | 32 | 5912 | |||
XM_011536621.2 | XP_011534923.1 | 31 | 5081 | |||
XM_011536622.1 | XP_011534924.1 | 23 | 4245 | |||
XM_017021148.1 | XP_016876637.1 | 31 | 5671 | |||
XM_017021149.1 | XP_016876638.1 | 31 | 5665 | |||
XM_017021151.1 | XP_016876640.1 | 31 | 5677 | |||
XM_017021152.1 | XP_016876641.1 | 31 | 5671 | |||
XM_017021153.1 | XP_016876642.1 | 31 | 5075 | |||
GenBank mRNA | AB032957.2 | - | 24 | 4466 | ||
AK024847.1 | - | 10 | 2149 | |||
AK095634.1 | - | 23 | 4072 | |||
AK125788.1 | - | 6 | 1427 | |||
AL110222.1 | - | 7 | 838 | |||
AY254380.1 | - | 32 | 6120 | |||
AY739714.1 | - | 8 | 1634 |