Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145804.2 | NP_665803.2 | 12 | 2856 | ||
GenBank mRNA | AK091560.1 | - | 9 | 1074 | ||
AK095632.1 | - | 12 | 2103 | |||
AK291744.1 | - | 12 | 2857 | |||
BC093968.1 | - | 12 | 1968 | 7939813 | ||
BX640841.1 | - | 4 | 557 |