Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015388.3 | NP_056203.2 | 7 | 882 | ||
XM_005248989.4 | XP_005249046.1 | 7 | 918 | |||
XM_005248990.2 | XP_005249047.1 | 7 | 923 | |||
XM_011514458.2 | XP_011512760.1 | 7 | 1025 | |||
GenBank mRNA | AF162672.1 | - | 7 | 797 | ||
AK000946.1 | - | 7 | 818 | |||
AK021433.1 | - | 794 | ||||
AK021655.1 | - | 7 | 762 | |||
AK022757.1 | - | 7 | 781 | |||
AK298150.1 | - | 7 | 918 | |||
AK308961.1 | - | 3 | 1298 | |||
AL050274.1 | - | 7 | 683 | |||
BC019297.1 | - | 7 | 831 | 2905449 | ||
CA308984.1 | - | 3 | 708 | |||
CR533541.1 | - | 6 | 594 |