Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF345 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242472.1 | NP_001229401.1 | 3 | 1656 | ||
NM_001242474.1 | NP_001229403.1 | 4 | 1801 | |||
NM_001242475.1 | NP_001229404.1 | 3 | 1636 | |||
NM_001242476.1 | NP_001229405.1 | 4 | 1731 | |||
NM_003419.4 | NP_003410.1 | 3 | 1823 | |||
XM_011526704.2 | XP_011525006.1 | 3 | 1782 | |||
XM_017026568.1 | XP_016882057.1 | 3 | 1980 | |||
XM_017026569.1 | XP_016882058.1 | 3 | 1636 | |||
XM_017026570.1 | XP_016882059.1 | 4 | 1747 | |||
XM_017026571.1 | XP_016882060.1 | 4 | 1746 | |||
XM_017026572.1 | XP_016882061.1 | 4 | 1850 | |||
XM_017026573.1 | XP_016882062.1 | 3 | 1875 | |||
GenBank mRNA | AK094146.1 | - | 2 | 2230 | ||
AK297516.1 | - | 3 | 994 | |||
AK315069.1 | - | 3 | 1656 | |||
BC032863.1 | - | 3 | 1821 | 5270008 | ||
X78933.1 | - | 3 | 1641 |