Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TECPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015395.2 | NP_056210.1 | 12 | 2085 | ||
XM_005250253.3 | XP_005250310.1 | 12 | 2057 | |||
XM_017011937.1 | XP_016867426.1 | 11 | 1967 | |||
XM_017011938.1 | XP_016867427.1 | 12 | 2069 | |||
GenBank mRNA | AB037779.1 | - | 10 | 1661 | ||
AK057048.1 | - | 11 | 1812 | |||
AK292996.1 | - | 12 | 1987 | |||
AL117495.1 | - | 1 | 983 | |||
BC053591.1 | - | 12 | 1990 | 5502429 |