Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TECPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015395.2 | NP_056210.1 | 18, 19 | 2919 | ||
XM_005250253.3 | XP_005250310.1 | 18, 19 | 2891 | |||
XM_017011937.1 | XP_016867426.1 | 17, 18 | 2801 | |||
GenBank mRNA | AB037779.1 | - | 16, 17 | 2495 | ||
AK027072.1 | - | 7, 8 | 822 | |||
AL117495.1 | - | 7, 8 | 1816 | |||
BC012529.2 | - | 6, 7 | 742 | |||
BC053591.1 | - | 18, 19 | 2824 | 5502429 |