Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079880.1 | NP_001073349.1 | 12, 13 | 2118 | ||
NM_001079881.1 | NP_001073350.1 | 12, 13 | 1982 | |||
NM_001079882.1 | NP_001073351.1 | 10, 11 | 1719 | |||
NM_016457.4 | NP_057541.2 | 11, 12 | 2397 | |||
XM_005258716.2 | XP_005258773.2 | 10, 11 | 1375 | |||
GenBank mRNA | AF309082.1 | - | 11, 12 | 1679 | ||
AK074673.1 | - | 10, 11 | 1358 | |||
AK075266.1 | - | 10, 11 | 1468 | |||
AK092896.1 | - | 11, 12 | 1769 | |||
AK095884.1 | - | 12, 13 | 1661 | |||
AK300339.1 | - | 12, 13 | 1932 | |||
AK303629.1 | - | 10, 11 | 1373 | |||
AL050147.1 | - | 5, 6 | 634 | |||
BC015472.1 | - | 2, 3 | 96 | |||
BC025307.1 | - | 7, 8 | 968 |