Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LETMD1 SIRNA Assays ›
Gene Symbol:
LETMD1
Gene Name:
LETM1 domain containing 1
Gene Aliases:
1110019O13Rik, HCCR-1, HCCR-2, HCCR1, HCRR-2
Chromosome Location:
Chr.12: 51048230 - 51068984 on Build GRCh38
UniGene ID:
Hs.655272
Species:
Human
siRNA ID
HSS119434
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LETMD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243689.1
NP_001230618.1
2
219
NM_001300765.1
NP_001287694.1
2
219
NM_015416.4
NP_056231.3
2
219
NR_045017.1
2
94
NR_045018.1
2
219
NR_045019.1
2
219
NR_045020.1
2
219
XM_006719335.1
XP_006719398.1
2
191
XM_006719336.1
XP_006719399.1
2
79
XM_006719339.1
XP_006719402.1
2
191
XM_006719340.1
XP_006719403.1
2
94
XM_011538161.2
XP_011536463.1
2
226
XM_011538162.2
XP_011536464.1
2
227
XM_011538163.2
XP_011536465.1
2
228
XM_011538164.2
XP_011536466.1
2
226
XM_011538165.2
XP_011536467.1
2
226
XM_017019151.1
XP_016874640.1
2
228
XM_017019152.1
XP_016874641.1
2
194
XM_017019153.1
XP_016874642.1
2
228
XM_017019154.1
XP_016874643.1
2
191
XM_017019155.1
XP_016874644.1
2
66
XM_017019156.1
XP_016874645.1
2
229
XM_017019157.1
XP_016874646.1
2
191
XM_017019158.1
XP_016874647.1
2
79
XM_017019159.1
XP_016874648.1
2
229
XM_017019160.1
XP_016874649.1
2
191
XM_017019161.1
XP_016874650.1
2
230
XM_017019162.1
XP_016874651.1
2
200
XM_017019163.1
XP_016874652.1
2
191
XM_017019164.1
XP_016874653.1
2
211
XM_017019165.1
XP_016874654.1
2
191
XM_017019166.1
XP_016874655.1
2
94
XM_017019167.1
XP_016874656.1
2
191
XM_017019168.1
XP_016874657.1
2
191
GenBank mRNA
AF195651.2
-
2
191
AF315598.1
-
2
55
AF329277.1
-
2
218
AK000639.1
-
2
206
AK057866.1
-
2
164
AK127540.1
-
2
180
AK225836.1
-
2
177
AK301898.1
-
2
179
AK301955.1
-
2
94
AK304707.1
-
2
191
AK315119.1
-
2
186
AL050286.1
-
1
199
AY259835.1
-
2
161
AY259836.1
-
2
161
BC064943.1
-
2
165
6009854
BM549678.1
-
191
BX343550.2
-
2
160
BX376670.2
-
2
160
DA682074.1
-
2
94
DC357852.1
-
2
219
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Process(es)
protein binding
ribosome binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0