Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNOT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256741.1 | NP_001243670.1 | 18 | 2360 | ||
NM_001256742.1 | NP_001243671.1 | 19 | 2390 | |||
NM_015442.2 | NP_056257.1 | 19 | 2441 | |||
NR_046352.1 | 17 | 2250 | ||||
XM_006713084.3 | XP_006713147.1 | 19 | 2394 | |||
XM_006713085.3 | XP_006713148.1 | 18 | 2361 | |||
XM_011533566.2 | XP_011531868.1 | 18 | 2316 | |||
XM_011533567.2 | XP_011531869.1 | 15 | 1658 | |||
XM_017006109.1 | XP_016861598.1 | 19 | 2438 | |||
XM_017006110.1 | XP_016861599.1 | 18 | 2405 | |||
XM_017006111.1 | XP_016861600.1 | 18 | 2402 | |||
XM_017006112.1 | XP_016861601.1 | 17 | 2324 | |||
GenBank mRNA | AK021695.1 | - | 16 | 1728 | ||
AK022576.1 | - | 19 | 2243 | |||
AK022952.1 | - | 19 | 2380 | |||
AK023227.1 | - | 19 | 2398 | |||
AK027026.1 | - | 16 | 1883 | |||
AK302196.1 | - | 19 | 2382 | |||
AK309670.1 | - | 17 | 2175 | |||
AL117639.1 | - | 8 | 692 | |||
BC002928.2 | - | 18 | 2262 | 3954507 | ||
BC002931.2 | - | 19 | 2349 | 3955575 | ||
BM979742.1 | - | 1 | 385 |