Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318520.1 | NP_001305449.1 | 46 | 6390 | ||
NM_173630.3 | NP_775901.3 | 47 | 6573 | |||
XM_006722434.3 | XP_006722497.1 | 47 | 6576 | |||
XM_011525902.2 | XP_011524204.1 | 46 | 6336 | |||
XM_011525903.2 | XP_011524205.1 | 45 | 6147 | |||
XM_011525906.2 | XP_011524208.1 | 37 | 5857 | |||
XM_017025693.1 | XP_016881182.1 | 46 | 6333 | |||
XM_017025694.1 | XP_016881183.1 | 44 | 6131 | |||
XM_017025695.1 | XP_016881184.1 | 40 | 5633 | |||
XM_017025696.1 | XP_016881185.1 | 47 | 6573 | |||
GenBank mRNA | AK126538.1 | - | 22 | 3018 | ||
AK128137.1 | - | 28 | 3944 | |||
AK129866.1 | - | 8 | 1058 | |||
AL117635.1 | - | 3 | 477 | |||
BC007359.2 | - | 3 | 428 | |||
BC026879.1 | - | 2 | 302 | |||
BC046222.2 | - | 12 | 1771 | |||
BC047602.1 | - | 39 | 5481 | |||
BC143723.1 | - | 46 | 6367 | |||
CR749799.1 | - | 41 | 5758 |