Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284423.1 | NP_001271352.1 | 10 | 1704 | ||
NM_001284424.1 | NP_001271353.1 | 10 | 1893 | |||
NM_001284425.1 | NP_001271354.1 | 9 | 1756 | |||
NM_001284426.1 | NP_001271355.1 | 8 | 1514 | |||
NM_015460.3 | NP_056275.2 | 10 | 1893 | |||
NR_104316.1 | 10 | 1893 | ||||
XM_011533575.1 | XP_011531877.1 | 10 | 1865 | |||
GenBank mRNA | AB083783.1 | - | 9 | 1606 | ||
AF396687.1 | - | 10 | 1742 | |||
AK125334.1 | - | 8 | 1380 | |||
AK294714.1 | - | 8 | 1504 | |||
AK312412.1 | - | 10 | 1703 | |||
AK316068.1 | - | 8 | 1514 | |||
BC092512.1 | - | 10 | 1892 | 3093499 | ||
BC109311.1 | - | 9 | 1584 | 40036096 | ||
BC109312.1 | - | 10 | 1721 | 40036097 |