Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEMIN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252156.1 | NP_001239085.1 | 11 | 1597 | ||
NM_015465.4 | NP_056280.2 | 11 | 1600 | |||
GenBank mRNA | AK022748.1 | - | 11 | 1547 | ||
AK074066.2 | - | 11 | 1572 | |||
AK315101.1 | - | 11 | 1585 | |||
AL117665.1 | - | 6 | 662 | |||
AY063750.1 | - | 11 | 1517 | |||
BC036894.1 | - | 11 | 1580 | |||
BC040931.1 | - | 11 | 1570 | |||
BC113614.1 | - | 11 | 1556 | 8322666 | ||
BC143724.1 | - | 11 | 1553 | 9052238 |