Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEMIN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252156.1 | NP_001239085.1 | 3 | 512 | ||
NM_015465.4 | NP_056280.2 | 3 | 512 | |||
GenBank mRNA | AK022748.1 | - | 3 | 459 | ||
AK074066.2 | - | 3 | 484 | |||
AK315101.1 | - | 3 | 497 | |||
AY063750.1 | - | 3 | 429 | |||
BC036894.1 | - | 3 | 492 | |||
BC040931.1 | - | 3 | 482 | |||
BC113614.1 | - | 3 | 468 | 8322666 | ||
BC143724.1 | - | 3 | 468 | 9052238 | ||
DA328736.1 | - | 3 | 512 | |||
DA400244.1 | - | 3 | 513 |