Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECTIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243286.1 | NP_001230215.1 | 6 | 1425 | ||
NM_015480.2 | NP_056295.1 | 6 | 1451 | |||
XM_011512663.1 | XP_011510965.1 | 7 | 1515 | |||
XM_017006124.1 | XP_016861613.1 | 7 | 1172 | |||
XM_017006125.1 | XP_016861614.1 | 6 | 1375 | |||
XM_017006127.1 | XP_016861616.1 | 6 | 1538 | |||
GenBank mRNA | AF282874.1 | - | 6 | 1192 | ||
AL050071.1 | - | 5 | 768 | |||
BC001336.1 | - | 4 | 459 | |||
BC017572.1 | - | 1 | 39 | |||
BC067808.1 | - | 6 | 1308 | 5285526 |