Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145795.1 | NP_001139267.1 | 7, 8 | 1994 | ||
NM_001145796.1 | NP_001139268.1 | 7, 8 | 1994 | |||
NM_001145797.1 | NP_001139269.1 | 7, 8 | 1994 | |||
NM_001145812.1 | NP_001139284.1 | 7, 8 | 2093 | |||
NM_001308293.1 | NP_001295222.1 | 9, 10 | 2250 | |||
NM_001308294.1 | NP_001295223.1 | 6, 7 | 929 | |||
NM_015503.2 | NP_056318.2 | 6, 7 | 3824 | |||
XM_017023114.1 | XP_016878603.1 | 6, 7 | 2413 | |||
XM_017023115.1 | XP_016878604.1 | 8, 9 | 2120 | |||
XM_017023116.1 | XP_016878605.1 | 7, 8 | 3478 | |||
XM_017023117.1 | XP_016878606.1 | 5, 6 | 2209 | |||
XM_017023118.1 | XP_016878607.1 | 6, 7 | 1100 | |||
XM_017023119.1 | XP_016878608.1 | 6, 7 | 929 | |||
XM_017023120.1 | XP_016878609.1 | 5, 6 | 763 | |||
GenBank mRNA | AB037720.1 | - | 6, 7 | 5013 | ||
AF227967.1 | - | 7, 8 | 1850 | |||
AF227968.1 | - | 7, 8 | 1850 | |||
AF227969.1 | - | 7, 8 | 1850 | |||
AK027488.1 | - | 6, 7 | 1159 | |||
AK055104.1 | - | 9, 10 | 2161 | |||
AK290332.1 | - | 7, 8 | 2093 | |||
AK297080.1 | - | 6, 7 | 880 | |||
AK298569.1 | - | 6, 7 | 979 | |||
AL049924.1 | - | 6, 7 | 1294 | |||
AL713760.1 | - | 6, 7 | 3824 | |||
BC010704.1 | - | 6, 7 | 1547 | 3866324 |