Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152268.3 | NP_689481.2 | 2 | 1511 | ||
GenBank mRNA | AK025585.1 | - | 1 | 1193 | ||
AK289679.1 | - | 2 | 1463 | |||
AL117473.1 | - | 1 | 1192 | |||
BC007956.2 | - | 1 | 1426 | 4301987 | ||
BC011758.2 | - | 2 | 1472 | 3530065 |