Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIPARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184717.1 | NP_001171646.1 | 2 | 673 | ||
NM_001184718.1 | NP_001171647.1 | 2 | 238 | |||
NM_015508.4 | NP_056323.2 | 2 | 588 | |||
GenBank mRNA | AK097785.1 | - | 2 | 417 | ||
AK291261.1 | - | 2 | 423 | |||
BC034397.1 | - | 2 | 238 | |||
BC050350.1 | - | 2 | 395 | 5264807 | ||
BX537965.1 | - | 2 | 415 | |||
CR749647.1 | - | 2 | 674 |