Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REXO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015523.3 | NP_056338.2 | 4 | 510 | ||
GenBank mRNA | AF151872.1 | - | 4 | 461 | ||
AK223200.1 | - | 4 | 425 | |||
AK312042.1 | - | 4 | 439 | |||
AL110239.1 | - | 4 | 444 | |||
BC105024.1 | - | 4 | 431 | 8144027 | ||
BC105026.1 | - | 4 | 431 | 8144029 | ||
BC107887.1 | - | 420 | 6663083 | |||
BC143701.1 | - | 4 | 434 | 9052215 | ||
CR533500.1 | - | 4 | 367 |