Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320356.1 | NP_001307285.1 | 8, 9 | 1076 | ||
NM_001320357.1 | NP_001307286.1 | 7, 8 | 966 | |||
NM_001320358.1 | NP_001307287.1 | 7, 8 | 836 | |||
NM_001320359.1 | NP_001307288.1 | 8, 9 | 992 | |||
NM_001320360.1 | NP_001307289.1 | 7, 8 | 1222 | |||
NM_001320361.1 | NP_001307290.1 | 8, 9 | 1206 | |||
NM_001320362.1 | NP_001307291.1 | 7, 8 | 786 | |||
NM_001320363.1 | NP_001307292.1 | 7, 8 | 916 | |||
NM_001320364.1 | NP_001307293.1 | 6, 7 | 845 | |||
NM_001320365.1 | NP_001307294.1 | 8, 9 | 1156 | |||
NM_015528.2 | NP_056343.1 | 7, 8 | 1352 | |||
XM_017024427.1 | XP_016879916.1 | 6, 7 | 1092 | |||
GenBank mRNA | AK025329.1 | - | 7, 8 | 1025 | ||
AL050060.1 | - | 5, 6 | 502 | |||
AL834284.1 | - | 6, 7 | 874 | |||
AY203930.1 | - | 6, 7 | 852 | |||
BC010139.2 | - | 6, 7 | 750 | 4128232 | ||
BC013222.1 | - | 6, 7 | 806 | |||
BI825561.1 | - | 7, 8 | 688 | |||
CR457340.1 | - | 6, 7 | 576 |