Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GORASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201428.1 | NP_001188357.1 | 4, 5 | 503 | ||
NM_015530.4 | NP_056345.3 | 4, 5 | 1258 | |||
XM_006712408.2 | XP_006712471.1 | 4, 5 | 581 | |||
GenBank mRNA | AK023082.1 | - | 4, 5 | 530 | ||
AK023201.1 | - | 4, 5 | 543 | |||
AK027349.1 | - | 4, 5 | 457 | |||
AK125761.1 | - | 4, 5 | 1258 | |||
AK225810.1 | - | 4, 5 | 540 | |||
AK296698.1 | - | 5, 6 | 567 | |||
AK298020.1 | - | 4, 5 | 537 | |||
AK298771.1 | - | 2, 3 | 124 | |||
AK315508.1 | - | 4, 5 | 559 | |||
AL117430.1 | - | 4, 5 | 469 | |||
BC007770.2 | - | 4, 5 | 492 | 4298251 | ||
DC399516.1 | - | 4, 5 | 505 |