Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SPATS2L SIRNA Assays ›
Gene Symbol:
SPATS2L
Gene Name:
spermatogenesis associated serine rich 2 like
Gene Aliases:
DNAPTP6, SGNP
Chromosome Location:
Chr.2: 200305881 - 200482263 on Build GRCh38
UniGene ID:
Hs.120323
Species:
Human
siRNA ID
HSS119663
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SPATS2L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001100422.1
NP_001093892.1
13
1737
NM_001100423.1
NP_001093893.1
13
1687
NM_001100424.1
NP_001093894.1
12
1530
NM_001282735.1
NP_001269664.1
13
1809
NM_001282743.1
NP_001269672.1
12
1748
NM_001282744.1
NP_001269673.1
13
1538
NM_015535.2
NP_056350.2
13
1677
XM_005246456.1
XP_005246513.1
14
1596
XM_005246457.1
XP_005246514.1
14
1859
XM_005246458.1
XP_005246515.1
12
1470
XM_005246459.1
XP_005246516.1
12
1277
XM_011510938.1
XP_011509240.1
14
1827
XM_011510939.1
XP_011509241.1
13
1540
XM_017003783.1
XP_016859272.1
14
1691
XM_017003784.1
XP_016859273.1
12
3805
XM_017003785.1
XP_016859274.1
11
1263
XM_017003786.1
XP_016859275.1
13
1389
XM_017003787.1
XP_016859276.1
13
1652
GenBank mRNA
AF193059.1
-
13
1623
AK002064.1
-
13
1475
AK090867.1
-
12
1470
AK092917.1
-
1
1044
AK290496.1
-
13
1797
AK299223.1
-
12
1450
AK300075.1
-
13
1540
AK314561.1
-
12
1238
AL110124.1
-
13
1485
AY450394.1
-
13
1485
BC018736.1
-
12
1223
4845423
Back To Top
More Information
Gene Ontology Categories:
Process(es)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0