Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM32A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014077.3 | NP_054796.1 | 4 | 362 | ||
XM_011527898.1 | XP_011526200.1 | 3 | 525 | |||
GenBank mRNA | AF151902.1 | - | 4 | 321 | ||
AK312058.1 | - | 4 | 345 | |||
AL050157.1 | - | 4 | 340 | |||
BC000639.1 | - | 4 | 340 | 3345441 | ||
BC004447.1 | - | 3 | 192 | |||
BC017286.1 | - | 4 | 323 | 4892079 | ||
BG749750.1 | - | 3 | 489 |