Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015542.3 | NP_056357.1 | 7 | 1807 | ||
NM_080599.2 | NP_542166.1 | 7 | 1977 | |||
XM_011519449.2 | XP_011517751.1 | 7 | 1855 | |||
XM_017016034.1 | XP_016871523.1 | 6 | 1463 | |||
GenBank mRNA | AB037829.1 | - | 6 | 2176 | ||
AF301013.1 | - | 7 | 1800 | |||
AF318574.1 | - | 7 | 1777 | |||
AK096191.1 | - | 8 | 1717 | |||
AK310925.1 | - | 7 | 1766 | |||
AY013249.1 | - | 6 | 1726 | |||
BC114964.1 | - | 7 | 1771 | 40082739 | ||
BC115737.1 | - | 7 | 1771 | 40082728 |