Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHGA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003735.2 | NP_003726.1 | 1 | 472 | ||
NM_032094.1 | NP_115265.1 | 1 | 472 | |||
GenBank mRNA | AB011160.1 | - | 1 | 445 | ||
AF152321.1 | - | 1 | 303 | |||
AF152506.1 | - | 1 | 303 | |||
AY358946.1 | - | 1 | 673 |