Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308147.1 | NP_001295076.1 | 16, 17 | 3263 | ||
XM_011536624.2 | XP_011534926.1 | 18, 19 | 3544 | |||
XM_011536625.2 | XP_011534927.1 | 18, 19 | 3417 | |||
XM_011536627.2 | XP_011534929.1 | 18, 19 | 3809 | |||
XM_011536628.2 | XP_011534930.1 | 18, 19 | 3995 | |||
XM_011536629.2 | XP_011534931.1 | 17, 18 | 3133 | |||
XM_017021154.1 | XP_016876643.1 | 17, 18 | 3410 | |||
XM_017021155.1 | XP_016876644.1 | 18, 19 | 3499 | |||
XM_017021156.1 | XP_016876645.1 | 18, 19 | 3567 | |||
XM_017021157.1 | XP_016876646.1 | 16, 17 | 3363 | |||
XM_017021158.1 | XP_016876647.1 | 16, 17 | 3002 | |||
GenBank mRNA | AB011171.1 | - | 1, 2 | 1361 | ||
AK074169.1 | - | 10, 11 | 3034 | |||
BC004298.1 | - | 4, 5 | 1073 | |||
BC011891.2 | - | 1, 2 | 1353 | |||
BC063554.1 | - | 2, 3 | 1444 | |||
BC073907.1 | - | 3, 4 | 1551 | |||
BC129952.1 | - | 10, 11 | 2755 | 40126555 | ||
BC129953.1 | - | 16, 17 | 3074 | 40126558 |