Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015550.3 | NP_056365.1 | 16 | 2279 | ||
NM_145320.2 | NP_663160.1 | 14 | 1859 | |||
NM_145321.2 | NP_663161.1 | 14 | 1844 | |||
NM_145322.2 | NP_663162.1 | 13 | 1751 | |||
NR_104111.1 | 15 | 2351 | ||||
NR_104112.1 | 14 | 2258 | ||||
XM_005249698.3 | XP_005249755.1 | 16 | 1999 | |||
XM_006715681.3 | XP_006715744.1 | 15 | 2173 | |||
XM_006715682.3 | XP_006715745.1 | 15 | 2177 | |||
XM_006715683.3 | XP_006715746.1 | 14 | 2084 | |||
XM_006715684.3 | XP_006715747.1 | 15 | 1936 | |||
XM_011515258.2 | XP_011513560.1 | 16 | 2076 | |||
XM_011515259.2 | XP_011513561.1 | 17 | 2573 | |||
XM_017011948.1 | XP_016867437.1 | 18 | 2436 | |||
XM_017011949.1 | XP_016867438.1 | 16 | 2465 | |||
GenBank mRNA | AB014604.2 | - | 16 | 2216 | ||
AF392444.1 | - | 16 | 2188 | |||
AF491781.1 | - | 15 | 1906 | |||
AF491782.1 | - | 14 | 1813 | |||
AF491783.1 | - | 14 | 1798 | |||
AF491784.1 | - | 13 | 1705 | |||
AF491785.1 | - | 15 | 2305 | |||
AF491786.1 | - | 14 | 2212 | |||
AL832293.1 | - | 13 | 1515 | |||
AY008372.1 | - | 16 | 2109 | |||
BC017731.1 | - | 16 | 2208 | 3909164 |