Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015550.3 | NP_056365.1 | 18 | 2462 | ||
NM_145320.2 | NP_663160.1 | 16 | 2042 | |||
NM_145321.2 | NP_663161.1 | 16 | 2027 | |||
NM_145322.2 | NP_663162.1 | 15 | 1934 | |||
NR_104111.1 | 17 | 2534 | ||||
NR_104112.1 | 16 | 2441 | ||||
XM_005249698.3 | XP_005249755.1 | 18 | 2182 | |||
XM_006715681.3 | XP_006715744.1 | 17 | 2356 | |||
XM_006715682.3 | XP_006715745.1 | 17 | 2360 | |||
XM_006715683.3 | XP_006715746.1 | 16 | 2267 | |||
XM_006715684.3 | XP_006715747.1 | 17 | 2119 | |||
XM_011515258.2 | XP_011513560.1 | 18 | 2259 | |||
XM_011515259.2 | XP_011513561.1 | 19 | 2756 | |||
XM_017011948.1 | XP_016867437.1 | 20 | 2619 | |||
XM_017011949.1 | XP_016867438.1 | 18 | 2648 | |||
GenBank mRNA | AB014604.2 | - | 18 | 2399 | ||
AF392444.1 | - | 18 | 2371 | |||
AF491781.1 | - | 17 | 2089 | |||
AF491782.1 | - | 16 | 1996 | |||
AF491783.1 | - | 16 | 1981 | |||
AF491784.1 | - | 15 | 1888 | |||
AF491785.1 | - | 17 | 2488 | |||
AF491786.1 | - | 16 | 2395 | |||
AL832293.1 | - | 15 | 1698 | |||
AY008372.1 | - | 18 | 2292 | |||
BC017731.1 | - | 18 | 2391 | 3909164 |