Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R16B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172735.2 | NP_001166206.1 | 9 | 1247 | ||
NM_015568.3 | NP_056383.1 | 10 | 1373 | |||
XM_011528768.2 | XP_011527070.1 | 9 | 1936 | |||
XM_011528769.2 | XP_011527071.1 | 8 | 954 | |||
XM_017027785.1 | XP_016883274.1 | 9 | 1268 | |||
GenBank mRNA | AB020630.2 | - | 10 | 1361 | ||
AB177855.1 | - | 8 | 874 | |||
AF362910.1 | - | 9 | 1224 | |||
BC131801.1 | - | 10 | 1330 | 40081818 | ||
BC152467.1 | - | 10 | 1361 | 8862603 |