Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DNM3 SIRNA Assays ›
Gene Symbol:
DNM3
Gene Name:
dynamin 3
Gene Aliases:
Dyna III
Chromosome Location:
Chr.1: 171841478 - 172418466 on Build GRCh38
UniGene ID:
Hs.654775
Species:
Human
siRNA ID
HSS119737
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DNM3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136127.2
NP_001129599.1
8
1248
NM_001278252.1
NP_001265181.1
8
1248
NM_015569.4
NP_056384.2
8
1248
XM_005245079.1
XP_005245136.1
8
1231
XM_005245080.1
XP_005245137.1
8
1231
XM_006711268.2
XP_006711331.1
8
1231
XM_017000976.1
XP_016856465.1
8
1231
XM_017000977.1
XP_016856466.1
8
1231
XM_017000978.1
XP_016856467.1
8
1231
XM_017000979.1
XP_016856468.1
8
1231
XM_017000980.1
XP_016856469.1
8
949
XM_017000981.1
XP_016856470.1
8
1231
XM_017000982.1
XP_016856471.1
8
1231
XM_017000983.1
XP_016856472.1
8
1231
XM_017000984.1
XP_016856473.1
8
1231
XM_017000985.1
XP_016856474.1
8
1231
XM_017000986.1
XP_016856475.1
8
1572
XM_017000987.1
XP_016856476.1
8
1103
XM_017000988.1
XP_016856477.1
8
1231
XM_017000989.1
XP_016856478.1
8
1231
XM_017000990.1
XP_016856479.1
8
1231
XM_017000991.1
XP_016856480.1
8
1231
XM_017000992.1
XP_016856481.1
8
1231
GenBank mRNA
AB020627.2
-
8
1199
AK056279.1
-
8
1228
AK307806.1
-
8
1186
AK308992.1
-
8
1197
AL136712.1
-
8
1226
BC064546.1
-
8
1089
5744324
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
synapse assembly
filopodium assembly
positive regulation of filopodium assembly
negative regulation of dendritic spine morphogenesis
GTPase activity
protein binding
GTP binding
type 1 metabotropic glutamate receptor binding
type 5 metabotropic glutamate receptor binding
nitric-oxide synthase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0