Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SENP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100409.2 | NP_001093879.1 | 7, 8 | 1691 | ||
NM_001304792.1 | NP_001291721.1 | 7, 8 | 1691 | |||
NM_015571.3 | NP_056386.2 | 8, 9 | 1712 | |||
XM_006715457.3 | XP_006715520.1 | 9, 10 | 1466 | |||
XM_011535746.2 | XP_011534048.1 | 7, 8 | 1264 | |||
XM_011535747.2 | XP_011534049.1 | 7, 8 | 1615 | |||
XM_017010719.1 | XP_016866208.1 | 8, 9 | 1354 | |||
XM_017010720.1 | XP_016866209.1 | 8, 9 | 670 | |||
XM_017010721.1 | XP_016866210.1 | 8, 9 | 678 | |||
XM_017010722.1 | XP_016866211.1 | 9, 10 | 1485 | |||
XM_017010723.1 | XP_016866212.1 | 9, 10 | 1462 | |||
XM_017010724.1 | XP_016866213.1 | 9, 10 | 1837 | |||
XM_017010725.1 | XP_016866214.1 | 6, 7 | 1597 | |||
GenBank mRNA | AB018340.2 | - | 8, 9 | 740 | ||
AF196304.1 | - | 8, 9 | 696 | |||
AF306508.1 | - | 8, 9 | 711 | |||
AF307849.1 | - | 8, 9 | 771 | |||
AK001406.1 | - | 5, 6 | 366 | |||
AK096455.1 | - | 7, 8 | 1273 | |||
AK126921.1 | - | 7, 8 | 1691 | |||
BC028583.1 | - | 7, 8 | 898 | 4838569 |