Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIGYF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103146.1 | NP_001096616.1 | 20 | 2490 | ||
NM_001103147.1 | NP_001096617.1 | 22 | 2623 | |||
NM_001103148.1 | NP_001096618.1 | 19 | 2406 | |||
NM_015575.3 | NP_056390.2 | 22 | 2630 | |||
NR_103492.1 | 19 | 2406 | ||||
GenBank mRNA | AB014542.2 | - | 22 | 2582 | ||
AK001739.1 | - | 22 | 2604 | |||
AK299418.1 | - | 21 | 2534 | |||
AK302998.1 | - | 14 | 1992 | |||
AY176045.1 | - | 22 | 2604 | |||
BC014047.2 | - | 7 | 814 | |||
BC043402.1 | - | 20 | 2425 | |||
BC136251.1 | - | 19 | 2361 | 9020238 | ||
BC143352.1 | - | 19 | 2361 | |||
BC146775.1 | - | 22 | 2582 | 9007224 | ||
BX537885.1 | - | 20 | 2425 | |||
BX538172.1 | - | 19 | 2374 | |||
BX538321.1 | - | 20 | 2446 | |||
BX647172.1 | - | 4 | 1328 | |||
BX647505.1 | - | 19 | 2237 |