Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIGYF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103146.1 | NP_001096616.1 | 23 | 3033 | ||
NM_001103147.1 | NP_001096617.1 | 25 | 3166 | |||
NM_001103148.1 | NP_001096618.1 | 22 | 2949 | |||
NM_015575.3 | NP_056390.2 | 25 | 3173 | |||
NR_103492.1 | 22 | 2949 | ||||
GenBank mRNA | AB014542.2 | - | 25 | 3125 | ||
AK001739.1 | - | 25 | 3147 | |||
AK027021.1 | - | 2 | 318 | |||
AK302998.1 | - | 17 | 2535 | |||
AY176045.1 | - | 25 | 3147 | |||
BC014047.2 | - | 10 | 1357 | |||
BC136251.1 | - | 22 | 2904 | 9020238 | ||
BC143352.1 | - | 22 | 2904 | |||
BC146775.1 | - | 25 | 3125 | 9007224 | ||
BX537885.1 | - | 23 | 2968 | |||
BX538172.1 | - | 22 | 2917 | |||
BX538321.1 | - | 23 | 2989 | |||
BX647172.1 | - | 7 | 1871 | |||
BX647505.1 | - | 22 | 2780 |