Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HACL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284413.1 | NP_001271342.1 | 2, 3 | 570 | ||
NM_001284415.1 | NP_001271344.1 | 2, 3 | 570 | |||
NM_001284416.1 | NP_001271345.1 | 2, 3 | 570 | |||
NM_012260.3 | NP_036392.2 | 2, 3 | 570 | |||
NR_104315.1 | 2, 3 | 570 | ||||
GenBank mRNA | AF161397.1 | - | 2, 3 | 190 | ||
AJ131753.1 | - | 2, 3 | 280 | |||
AK301546.1 | - | 2, 3 | 287 | |||
AK301990.1 | - | 2, 3 | 305 | |||
AK302086.1 | - | 2, 3 | 287 | |||
BC001627.1 | - | 2, 3 | 288 | 3538159 | ||
BC007440.2 | - | 2, 3 | 275 | |||
DA609383.1 | - | 2, 3 | 287 | |||
DB049608.1 | - | 2, 3 | 341 |