Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251962.1 | NP_001238891.1 | 15 | 2815 | ||
NM_001251963.1 | NP_001238892.1 | 15 | 2677 | |||
NM_015595.3 | NP_056410.3 | 15 | 2742 | |||
XM_011512672.1 | XP_011510974.1 | 14 | 2652 | |||
XM_017006160.1 | XP_016861649.1 | 14 | 1484 | |||
GenBank mRNA | AF415175.1 | - | 15 | 2697 | ||
AF415176.1 | - | 5 | 667 | |||
AK022655.1 | - | 9 | 1053 | |||
AK022884.1 | - | 12 | 1329 | |||
AK123040.1 | - | 15 | 2621 | |||
AK127282.1 | - | 15 | 2815 | |||
AL117429.1 | - | 5 | 553 | |||
AY552599.1 | - | 14 | 2531 | |||
BC016628.1 | - | 9 | 961 | |||
BC078655.1 | - | 15 | 2683 | 4374088 |