Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015596.1 | NP_056411.1 | 4 | 602 | ||
GenBank mRNA | AB108823.1 | - | 2 | 103 | ||
AB108824.1 | - | 4 | 340 | |||
AB451465.1 | - | 4 | 559 | |||
AY923171.1 | - | 4 | 391 | |||
AY923172.1 | - | 5 | 958 | |||
AY923173.1 | - | 5 | 667 | |||
AY923174.1 | - | 4 | 874 | |||
AY923175.1 | - | 4 | 583 | |||
BC069334.1 | - | 4 | 603 | 7262205 | ||
BC069543.1 | - | 4 | 602 | 7262217 |