Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278185.1 | NP_001265114.1 | 19 | 2388 | ||
NM_001278186.1 | NP_001265115.1 | 16 | 2112 | |||
NM_015601.3 | NP_056416.2 | 18 | 2364 | |||
NM_022079.2 | NP_071362.1 | 19 | 2388 | |||
XM_011539592.2 | XP_011537894.1 | 20 | 2462 | |||
XM_011539593.2 | XP_011537895.1 | 19 | 2438 | |||
XM_011539594.2 | XP_011537896.1 | 21 | 2548 | |||
XM_011539595.2 | XP_011537897.1 | 21 | 2598 | |||
XM_011539596.2 | XP_011537898.1 | 20 | 2476 | |||
XM_011539597.2 | XP_011537899.1 | 18 | 2099 | |||
XM_017016041.1 | XP_016871530.1 | 17 | 2027 | |||
GenBank mRNA | AB046813.1 | - | 16 | 2242 | ||
AK027835.1 | - | 9 | 931 | |||
AK292809.1 | - | 18 | 2244 | |||
AL096715.1 | - | 1 | 1761 | |||
AY221963.1 | - | 19 | 2325 | |||
AY650032.1 | - | 16 | 2049 | |||
AY650033.1 | - | 19 | 2325 | |||
BC015043.1 | - | 16 | 2015 | |||
BC039600.1 | - | 18 | 2274 | 5212048 | ||
BX537563.1 | - | 18 | 4851 |