Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR1AIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267578.1 | NP_001254507.1 | 10 | 1933 | ||
NM_015602.3 | NP_056417.2 | 10 | 1930 | |||
XM_011509403.1 | XP_011507705.1 | 11 | 1978 | |||
XM_011509404.1 | XP_011507706.1 | 11 | 1975 | |||
GenBank mRNA | AK001780.1 | - | 8 | 1318 | ||
AK021613.1 | - | 10 | 1211 | |||
AK023204.1 | - | 10 | 1161 | |||
AK291495.1 | - | 10 | 1118 | |||
AL050126.1 | - | 1 | 261 | |||
BC023247.1 | - | 10 | 1172 | 3458117 | ||
BX369105.2 | - | 4 | 673 |